1-Cyclopentylethanone 98%

Supplier: Thermo Scientific Chemicals
Warning

H64744-03 H64744-06 H64744-MD
AAH64744-03EA 78.7 USD
AAH64744-03 AAH64744-06 AAH64744-MD
1-Cyclopentylethanone 98%
1-Cyclopentylethanone

Purity: 98% CAS No.: 6004-60-0 Molecular Formula: C7H12O Formula Weight: 112.17 Physical Form: Liquid Density: 0.913 Refractive Index: 1.4435 Appearance: Colorless Boiling Point: 151-156° Beilstein Reference: 1853483 MDL No.: MFCD00060799

Formula: C₇H₁₂O
Boiling Pt: 151…156 °C
Density: 0.913
Storage Temperature: Ambient
MDL Number: MFCD00060799
CAS Number: 6004-60-0

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR