6-Chloro-5-fluoropicolinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H64734-03 H64734-06
AAH64734-03EA 57.3 USD
AAH64734-03 AAH64734-06 AAH64734-MD
6-Chloro-5-fluoropicolinic acid 98%
6-Chloro-5-fluoropicolinic acid

Purity: 98% CAS No.: 860296-24-8 Molecular Formula: C6H3ClFNO2 Formula Weight: 175.54 Physical Form: Solid Appearance: Off-white Beilstein Reference: 10063182 MDL No.: MFCD13185819

Formula: C₆H₃ClFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185819
CAS Number: 860296-24-8

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR