Methyl-4-chloroacetoacetate 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methyl 4-chloro-3-oxobutanoate
H64709-14
H64709-22
AAH64709-14EA
49.8
USD
AAH64709-14
AAH64709-22
Methyl-4-chloroacetoacetate 97+%
Methyl-4-chloroacetoacetate
Purity: 97+%
CAS No.: 32807-28-6
Molecular Formula: C5H7ClO3
Formula Weight: 150.56
Flash Point: 102°(216°F)
Physical Form: Liquid
Density: 1.305
Refractive Index: 1.4560
Appearance: Colorless
Boiling Point: 85°/4mm
Beilstein Reference: 1759939
UN No.: 2922
MDL No.: MFCD00000938
Formula:
C₅H₇ClO₃ MW: 150.56 g/mol Boiling Pt: 85 °C/4 mm Density: 1.305 Flash Pt: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000938 CAS Number: 32807-28-6 EINECS: 251-230-5 UN: 2922 ADR: 8,III |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...