tert-Butyl hydrogen succinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Succinic acid mono-tert-butyl ester, 4-(tert-Butoxy)-4-oxobutanoic acid, tert-Butyl Hydrogen Succinate, Mono-tert-butyl Succinate
H64706-03
H64706-06
AAH64706-03EA
36.7
USD
AAH64706-03
AAH64706-06
tert-Butyl hydrogen succinate 97%
tert-Butyl hydrogen succinate
Purity: 97%
CAS No.: 15026-17-2
Molecular Formula: C8H14O4
Formula Weight: 174.20
Flash Point:
Formula:
C₈H₁₄O₄ MW: 174.196 g/mol Melting Pt: 51…54 °C (lit.) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00273441 CAS Number: 15026-17-2 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...