4-(9-Carbazolyl)benzeneboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(9H-9-carbozale)phenylboronic acid, 4-(9H-Carbazol-9-yl)phenylboronic acid, (4-(9H-Carbazol-9-yl)phenyl)boronic acid, 4-(9H-Carbazol-9-yl)benzeneboronic acid
H64703-03
H64703-06
AAH64703-03EA
80.2
USD
AAH64703-03
AAH64703-06
AAH64703-MD
4-(9-Carbazolyl)benzeneboronic acid 98%
4-(9-Carbazolyl)benzeneboronic acid
Purity: 98%
CAS No.: 419536-33-7
Molecular Formula: C18H14BNO2
Formula Weight: 287.13
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 10565612
MDL No.: MFCD13176534
Formula:
C₁₈H₁₄BNO₂ MW: 287.125 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13176534 CAS Number: 419536-33-7 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...