2-Bromo-6-fluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Purity: 98%
CAS No.: 360575-28-6
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₇H₄BrFO Boiling Pt: 100…102 °C/8 mm Melting Pt: 43…47 °C Density: 1.70 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03407341 CAS Number: 360575-28-6 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...