Methyl-3-fluoro-4-nitrobenzoate 97%

Supplier: Thermo Scientific Chemicals
Warning

H64688-06 H64688-14
AAH64688-06EA 61.5 USD
AAH64688-06 AAH64688-14 AAH64688-22
Methyl-3-fluoro-4-nitrobenzoate 97%
Methyl-3-fluoro-4-nitrobenzoate

Purity: 97% CAS No.: 185629-31-6 Molecular Formula: C8H6FNO4 Formula Weight: 199.14 Physical Form: Solid Beilstein Reference: 10657755 MDL No.: MFCD08444027

Formula: C₈H₆FNO₄
Storage Temperature: Ambient
MDL Number: MFCD08444027
CAS Number: 185629-31-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR