5-Bromo-2-hydroxy-4-picoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64674-06
H64674-14
AAH64674-06EA
54.2
USD
AAH64674-06
AAH64674-14
5-Bromo-2-hydroxy-4-picoline 97%
5-Bromo-2-hydroxy-4-picoline
Purity: 97%
CAS No.: 164513-38-6
Molecular Formula: C6H6BrNO
Formula Weight: 188.02
Physical Form: Solid
Appearance: Off-white
Melting Point: 198-202°
Beilstein Reference: 6798531
MDL No.: MFCD03427664
Formula:
C₆H₆BrNO Melting Pt: 198…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD03427664 CAS Number: 164513-38-6 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...