4,4'-Dibromotriphenylamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N-Bis(p-bromophenyl)aniline

H64666-03 H64666-06
AAH64666-03EA 50.9 USD
AAH64666-03 AAH64666-06 AAH64666-MD
4,4'-Dibromotriphenylamine 98%
4,4'-Dibromotriphenylamine

Purity: 98% CAS No.: 81090-53-1 Molecular Formula: C18H13Br2N Formula Weight: 403.12 Flash Point: 110°(230°F) Physical Form: Solid Appearance: Off-white Melting Point: 85° Beilstein Reference: 7094354 MDL No.: MFCD00060104

Formula: C₁₈H₁₃Br₂N
MW: 403.12 g/mol
Melting Pt: 85 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060104
CAS Number: 81090-53-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR