2-Bromo-6-hydrazinylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-Bromo-6-hydrazinopyridine
H64664-06
H64664-14
AAH64664-03EA
24.87
USD
AAH64664-03
AAH64664-06
AAH64664-14
2-Bromo-6-hydrazinylpyridine 98%
2-Bromo-6-hydrazinylpyridine
Purity: 98%
CAS No.: 26944-71-8
Molecular Formula: C5H6BrN3
Formula Weight: 188.03
Physical Form: Solid
Appearance: Pale brown
Melting Point: 117-118°
Beilstein Reference: 386966
UN No.: 2811
MDL No.: MFCD09743754
Formula:
C₅H₆BrN₃ MW: 188.028 g/mol Melting Pt: 117…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD09743754 CAS Number: 26944-71-8 UN: 2811 ADR: "6.1",III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...