2-Bromo-1-(o-tolyl)ethanone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromo-1-(o-tolyl)ethan-1-one, 2-Bromo-2'-methylacetophenone

H64651-03 H64651-06
AAH64651-03EA 91 USD
AAH64651-03 AAH64651-06 AAH64651-MD
2-Bromo-1-(o-tolyl)ethanone 98%
2-Bromo-1-(o-tolyl)ethanone

Purity: 98% CAS No.: 51012-65-8 Molecular Formula: C9H9BrO Formula Weight: 213.07 Boiling Point: 81-83° Beilstein Reference: 508116 MDL No.: MFCD04038955

Formula: C₉H₉BrO
MW: 213.074 g/mol
Boiling Pt: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD04038955
CAS Number: 51012-65-8

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR