5-Bromo-2-iodobenzoic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64643-06
H64643-14
AAH64643-06EA
48.3
USD
AAH64643-06
AAH64643-14
5-Bromo-2-iodobenzoic acid 97%
5-Bromo-2-iodobenzoic acid
Purity: 97%
CAS No.: 21740-00-1
Molecular Formula: C7H4BrIO2
Formula Weight: 326.92
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 158-163°
Beilstein Reference: 3247380
MDL No.: MFCD00079717
Formula:
C₇H₄BrIO₂ Melting Pt: 158…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079717 CAS Number: 21740-00-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...