2-Amino-6-chloro-3-nitropyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine
H64634-14
H64634-22
AAH64634-14EA
78.7
USD
AAH64634-14
AAH64634-22
2-Amino-6-chloro-3-nitropyridine 98%
2-Amino-6-chloro-3-nitropyridine
Purity: 98%
CAS No.: 27048-04-0
Molecular Formula: C5H4ClN3O2
Formula Weight: 173.56
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Appearance: Yellow
Melting Point: 192-196°
Beilstein Reference: 474607
UN No.: 2811
MDL No.: MFCD00044350
Formula:
C₅H₄ClN₃O₂ MW: 173.56 g/mol Melting Pt: 192…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00044350 CAS Number: 27048-04-0 EINECS: 248-188-5 ADR: 6.1,III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...