3-Fluoro-4-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H64603-03 H64603-06
AAH64603-03EA 101 USD
AAH64603-03 AAH64603-06 AAH64603-MD
3-Fluoro-4-methoxypyridine 97%
3-Fluoro-4-methoxypyridine

Purity: 98% CAS No.: 1060805-03-9 Molecular Formula: C6H6FNO Formula Weight: 127.12 Physical Form: Solid Appearance: Off-white Beilstein Reference: 19561531 MDL No.: MFCD13185605

Formula: C₆H₆FNO
Storage Temperature: Ambient
MDL Number: MFCD13185605
CAS Number: 1060805-03-9

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR