5-Bromo-2,3-difluoropyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

H64566-03 H64566-06 H64566-14
AAH64566-03EA 38.7 USD
AAH64566-03 AAH64566-06 AAH64566-14
5-Bromo-2,3-difluoropyridine 97%
5-Bromo-2,3-difluoropyridine

Purity: 97% CAS No.: 89402-44-8 Molecular Formula: C5H2BrF2N Formula Weight: 193.98 Physical Form: Liquid Appearance: Colorless to pale yellow Beilstein Reference: 13412561 MDL No.: MFCD06659525

Formula: C5H2BrF2N
MW: 193.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659525
CAS Number: 89402-44-8

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR