4-Chloro-2-fluoropyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

H64557-03 H64557-06
AAH64557-03EA 170 USD
AAH64557-03 AAH64557-06 AAH64557-MD
4-Chloro-2-fluoropyridine 95%
4-Chloro-2-fluoropyridine

Purity: 95% CAS No.: 34941-92-9 Molecular Formula: C5H3ClFN Formula Weight: 131.53 Physical Form: Solid Beilstein Reference: 1634727 MDL No.: MFCD04112503

Formula: C5H3ClFN
Storage Temperature: Ambient
MDL Number: MFCD04112503
CAS Number: 34941-92-9

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR