1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 7-Deazahypoxanthine

H64537-06 H64537-14
AAH64537-03EA 21.59 USD
AAH64537-03 AAH64537-06 AAH64537-14
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one 97%
1H-Pyrrolo[2,3-d]pyrimidin-4(7H)-one

Purity: 97% CAS No.: 3680-71-5 Molecular Formula: C6H5N3O Formula Weight: 135.13 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Moisture and Light Sensitive Appearance: Brown Melting Point: 345-350° Beilstein Reference: 607869 MDL No.: MFCD08234923

Formula: C6H5N3O
MW: 135.13 g/mol
Melting Pt: 345…350 °C
Storage Temperature: Ambient
MDL Number: MFCD08234923
CAS Number: 3680-71-5

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR