9,9'-Spirobi[9H-fluorene]-2-boronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 9,9'-Spirobifluorene-2-boronic acid

H64525-03 H64525-06
AAH64525-03EA 77.8 USD
AAH64525-03 AAH64525-06 AAH64525-MD
9,9'-Spirobi[9H-fluorene]-2-boronic acid 98%
9,9'-Spirobi[9H-fluorene]-2-boronic acid

Purity: 98% CAS No.: 236389-21-2 Molecular Formula: C25H17BO2 Formula Weight: 360.22 Physical Form: Powder Appearance: Off-white Beilstein Reference: 10478712 MDL No.: MFCD13248568

Formula: C₂₅H₁₇BO₂
MW: 360.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13248568
CAS Number: 236389-21-2

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR