Methyl-4-methoxypicolinate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 4-methoxypyridine-2-carboxylate

H64519-06 H64519-14
AAH64519-03EA 19.95 USD
AAH64519-03 AAH64519-06 AAH64519-14
Methyl-4-methoxypicolinate 98%
Methyl 4-methoxypicolinate

Purity: 98% CAS No.: 29681-43-4 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Physical Form: Solid Appearance: White to pale yellow Melting Point: 47-50° Beilstein Reference: 473379 MDL No.: MFCD00956223

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: 47…50 °C
Storage Temperature: Ambient
MDL Number: MFCD00956223
CAS Number: 29681-43-4

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR