N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine, N4,N4'-Di(naphthalen-1-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
H64502-03
H64502-06
H64502-MD
AAH64502-03EA
108
USD
AAH64502-03
AAH64502-06
AAH64502-MD
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 98%
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine
Purity: 98%
CAS No.: 123847-85-8
Molecular Formula: C44H32N2
Formula Weight: 588.75
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 279-283°
Beilstein Reference: 8022378
MDL No.: MFCD03093246
Formula:
C₄₄H₃₂N₂ MW: 588.75 g/mol Melting Pt: 279…283 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093246 CAS Number: 123847-85-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...