tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 98%

Supplier: Thermo Scientific Chemicals

H64468-03 H64468-06 H64468-MD
AAH64468-03EA 65.7 USD
AAH64468-03 AAH64468-06 AAH64468-MD
tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride 98%
tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

Purity: 99% CAS No.: 77497-74-6 Molecular Formula: C14H19NO2.HCl Formula Weight: 269.77 Storage Temperature: 2°C to 8°C Physical Form: Solid Beilstein Reference: 5744730 MDL No.: MFCD04114841

Formula: C₁₄H₁₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD04114841
CAS Number: 77497-74-6

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR