Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate, Methyl 3,4-dihydro-3-oxo-2H-1,4-benzothiazine-6-carboxylate

H64467-06 H64467-14
AAH64467-03EA 47.85 USD
AAH64467-03 AAH64467-06 AAH64467-14
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate 97%
Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate

Purity: 97% CAS No.: 188614-01-9 Molecular Formula: C10H9NO3S Formula Weight: 223.25 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 97-99° Beilstein Reference: 14311523 MDL No.: MFCD00449104

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00449104
CAS Number: 188614-01-9

Order Now

Specification Test Results

Assay Minimum 97

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR