1-Aminocyclobutanecarboxylic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H64459-03 H64459-06
AAH64459-03EA 48.4 USD
AAH64459-03 AAH64459-06
1-Aminocyclobutanecarboxylic acid 95%
1-Aminocyclobutanecarboxylic acid

Purity: 95% CAS No.: 22264-50-2 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Physical Form: Powder Appearance: White Melting Point: 261° dec. Beilstein Reference: 2802253 MDL No.: MFCD00661068

Formula: C₅H₉NO₂
Melting Pt: 261 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00661068
CAS Number: 22264-50-2

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR