6-Azaindole ≥97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64458-03
H64458-06
AAH64458-03EA
66.4
USD
AAH64458-03
AAH64458-06
AAH64458-MD
6-Azaindole ≥97%
6-Azaindole
Purity: 98+%
CAS No.: 271-29-4
Molecular Formula: C7H6N2
Formula Weight: 118.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Air and Light Sensitive
Beilstein Reference: 109685
MDL No.: MFCD01686899
Formula:
C₇H₆N₂ MW: 118.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01686899 CAS Number: 271-29-4 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...