3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,6-Dihydro-2H-pyran-4-boronic acid Pinacol ester, 2-(3,6-Dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyran, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran

H64452-03 H64452-06
AAH64452-03EA 177 USD
AAH64452-03 AAH64452-06 AAH64452-MD
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester 98%
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester

Purity: 98% CAS No.: 287944-16-5 Molecular Formula: C11H19BO3 Formula Weight: 210.08 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Moisture Sensitive Appearance: White to off-white Melting Point: 63-67° Beilstein Reference: 8548328 MDL No.: MFCD11052631

Formula: C₁₁H₁₉BO₃
Melting Pt: 63…67 °C
Storage Temperature: Ambient
MDL Number: MFCD11052631
CAS Number: 287944-16-5

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR