2-Bromo-6-fluoro-4-methylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H64449-03 H64449-06
AAH64449-03EA 148 USD
AAH64449-03 AAH64449-06 AAH64449-MD
2-Bromo-6-fluoro-4-methylpyridine 98%
2-Bromo-6-fluoro-4-methylpyridine

Purity: 98% CAS No.: 180608-37-1 Molecular Formula: C6H5BrFN Formula Weight: 190.02 Physical Form: Liquid Appearance: Colorless to pale yellow Beilstein Reference: 13574124 MDL No.: MFCD09839269

Formula: C₆H₅BrFN
MW: 190.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839269
CAS Number: 180608-37-1

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR