4-methoxy-1-naphthoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64432-03
H64432-06
H64432-14
AAH64432-03EA
48
USD
AAH64432-03
AAH64432-06
AAH64432-14
4-methoxy-1-naphthoic acid 98%
4-methoxy-1-naphthoic acid
Purity: 98%
CAS No.: 13041-62-8
Molecular Formula: C12H10O3
Formula Weight: 202.21
Physical Form: Crystalline solid
Appearance: Off-white
Melting Point: 243-247°
Beilstein Reference: 2099800
MDL No.: MFCD02575372
Formula:
C₁₂H₁₀O₃ Melting Pt: 243…247 °C Storage Temperature: Ambient |
MDL Number:
MFCD02575372 CAS Number: 13041-62-8 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...