2-Bromoisophthalaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H64423-03 H64423-06
AAH64423-03EA 40.1 USD
AAH64423-03 AAH64423-06 AAH64423-MD
2-Bromoisophthalaldehyde 97%
2-Bromoisophthalaldehyde

Purity: 97% CAS No.: 79839-49-9 Molecular Formula: C8H5BrO2 Formula Weight: 213.03 Physical Form: Solid Melting Point: 139-144° Beilstein Reference: 4740331 MDL No.: MFCD08461034

Formula: C₈H₅BrO₂
MW: 213.03 g/mol
Melting Pt: 139…144 °C
Storage Temperature: Ambient
MDL Number: MFCD08461034
CAS Number: 79839-49-9

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR