2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64394-03
H64394-06
H64394-14
AAH64394-03EA
32
USD
AAH64394-03
AAH64394-06
AAH64394-14
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 95%
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 72587-15-6
Molecular Formula: C6H2ClF3N2O2
Formula Weight: 226.54
Physical Form: Liquid
Appearance: Colorless to pale yellow
Beilstein Reference: 7586778
MDL No.: MFCD06255171
Formula:
C6H2ClF3N2O2 MW: 226.54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06255171 CAS Number: 72587-15-6 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...