4-Bromo-2,6-difluorobenzotrifluoride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-1,3-difluoro-2-(trifluoromethyl)benzene
H64386-03
H64386-06
AAH64386-03EA
62.3
USD
AAH64386-03
AAH64386-06
AAH64386-MD
4-Bromo-2,6-difluorobenzotrifluoride 98%
4-Bromo-2,6-difluorobenzotrifluoride
Purity: 98%
CAS No.: 156243-64-0
Molecular Formula: C7H2BrF5
Formula Weight: 260.99
Physical Form: Liquid
Appearance: Colorless
Beilstein Reference: 13252396
MDL No.: MFCD01631616
Formula:
C₇H₂BrF₅ MW: 260.99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01631616 CAS Number: 156243-64-0 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...