Methyl-2-amino-4,5-difluorobenzoate 98%

Supplier: Thermo Scientific Chemicals
Warning

H64385-06 H64385-14 H64385-22
AAH64385-06EA 29.7 USD
AAH64385-06 AAH64385-14 AAH64385-22
Methyl-2-amino-4,5-difluorobenzoate 98%
Methyl-2-amino-4,5-difluorobenzoate

Purity: 98% CAS No.: 207346-42-7 Molecular Formula: C8H7F2NO2 Formula Weight: 187.15 Physical Form: Solid Appearance: White Beilstein Reference: 2834936 MDL No.: MFCD09264511

Formula: C8H7F2NO2
Storage Temperature: Ambient
MDL Number: MFCD09264511
CAS Number: 207346-42-7

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR