4-Bromo-3-chloro-2-fluoropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H64362-03 H64362-06 H64362-MD
AAH64362-03EA 77.2 USD
AAH64362-03 AAH64362-06 AAH64362-MD
4-Bromo-3-chloro-2-fluoropyridine 98%
4-Bromo-3-chloro-2-fluoropyridine

Purity: 98% CAS No.: 1017793-21-3 Molecular Formula: C5H2BrClFN Formula Weight: 210.43 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD11040607

Formula: C₅H₂BrClFN
MW: 210.43 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040607
CAS Number: 1017793-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR