2-Amino-4-bromopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromopyrimidin-2-amine

H64358-03 H64358-MD
AAH64358-03EA 318 USD
AAH64358-03 AAH64358-MD
2-Amino-4-bromopyrimidine 98%
2-Amino-4-bromopyrimidine

Purity: 98% CAS No.: 343926-69-2 Molecular Formula: C4H4BrN3 Formula Weight: 174.00 Physical Form: Powder Appearance: White Beilstein Reference: 111375 MDL No.: MFCD09999168

Formula: C₄H₄BrN₃
MW: 174.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999168
CAS Number: 343926-69-2

Order Now

Specification Test Results

Assay Minimum 98

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR