4-Bromo-4'-iodobiphenyl 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-4'-iodo-1,1'-biphenyl, 1-Bromo-4-(4-iodophenyl)benzene
H64355-14
H64355-22
AAH64355-14EA
102
USD
AAH64355-14
AAH64355-22
4-Bromo-4'-iodobiphenyl 98%
4-Bromo-4'-iodobiphenyl
Purity: 98%
CAS No.: 105946-82-5
Molecular Formula: C12H8BrI
Formula Weight: 359.00
Physical Form: Solid
Sensitivity: Light Sensitive
Melting Point: 175-179° lit.
Beilstein Reference: 2518914
UN No.: 3152
MDL No.: MFCD17676204
Formula:
C₁₂H₈BrI MW: 359.00 g/mol Melting Pt: 175…179 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD17676204 CAS Number: 105946-82-5 UN: 3152 ADR: "9",II |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...