β-Alanine-tert-butylester hydrochloride 95%

Supplier: Thermo Scientific Chemicals

H64346-06 H64346-14 H64346-22
AAH64346-06EA 69 USD
AAH64346-06 AAH64346-14 AAH64346-22
β-Alanine-tert-butylester hydrochloride 95%
beta-Alanine-tert-butylester hydrochloride

Purity: 95% CAS No.: 58620-93-2 Molecular Formula: C7H15NO2.HCl Formula Weight: 181.66 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 167-169° Beilstein Reference: 6660122 MDL No.: MFCD00058258

Formula: C₇H₁₆ClNO₂
Melting Pt: 167…169 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00058258
CAS Number: 58620-93-2

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR