3-Bromo-6-chloro-2-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64333-06
H64333-14
AAH64333-06EA
28
USD
AAH64333-06
AAH64333-14
3-Bromo-6-chloro-2-methylpyridine 98%
3-Bromo-6-chloro-2-methylpyridine
Purity: 98%
CAS No.: 132606-40-7
Molecular Formula: C6H5BrClN
Formula Weight: 206.47
Physical Form: Liquid
Melting Point: 46-47°
Boiling Point: 80-84°/2mm lit.
Beilstein Reference: 4243997
MDL No.: MFCD07375117
Formula:
C₆H₅BrClN Boiling Pt: 80…84 °C/2 mm (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD07375117 CAS Number: 132606-40-7 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...