Methyl-4-amino-3-bromobenzoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 4-amino-3-bromobenzoate
H64303-03
H64303-06
AAH64303-03EA
43.8
USD
AAH64303-03
AAH64303-06
Methyl-4-amino-3-bromobenzoate 97%
Methyl 4-amino-3-bromobenzoate
Purity: 97%
CAS No.: 106896-49-5
Molecular Formula: C8H8BrNO2
Formula Weight: 230.06
Physical Form: Solid
Melting Point: 105-109°
Beilstein Reference: 4905039
MDL No.: MFCD01861385
Formula:
C₈H₈BrNO₂ MW: 230.06 g/mol Melting Pt: 105…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861385 CAS Number: 106896-49-5 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...