2-Bromo-5-nitro-6-picoline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
6-Bromo-2-methyl-3-nitropyridine, 2-Bromo-6-methyl-5-nitropyridine
H64299-06
H64299-14
AAH64299-06EA
42.5
USD
AAH64299-06
AAH64299-14
2-Bromo-5-nitro-6-picoline 97%
2-Bromo-5-nitro-6-picoline
Purity: 97%
CAS No.: 22282-96-8
Molecular Formula: C6H5BrN2O2
Formula Weight: 217.02
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 67-72°
Beilstein Reference: 135841
MDL No.: MFCD03095219
Formula:
C₆H₅BrN₂O₂ MW: 217.02 Melting Pt: 67…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095219 CAS Number: 22282-96-8 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...