Ethyl-3-amino-3-iminopropanoate hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 2-amidinoacetate hydrochloride
H64288-06
H64288-14
AAH64288-03EA
29.08
USD
AAH64288-03
AAH64288-06
AAH64288-14
Ethyl-3-amino-3-iminopropanoate hydrochloride 95%
Ethyl-3-amino-3-iminopropanoate hydrochloride
Purity: 95%
CAS No.: 57508-48-2
Molecular Formula: C5H10N2O2.HCl
Formula Weight: 166.60
Physical Form: Solid
Melting Point: 105-108°
Beilstein Reference: 3694030
MDL No.: MFCD06797615
Formula:
C5H11ClN2O2 MW: 166.60 g/mol Melting Pt: 105…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797615 CAS Number: 57508-48-2 |
Specification Test Results
Assay Minimum | 95 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...