4-Fluoro-2-methylpyridine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64284-03
H64284-06
AAH64284-03EA
75.2
USD
AAH64284-03
AAH64284-06
AAH64284-MD
4-Fluoro-2-methylpyridine 96%
4-Fluoro-2-methylpyridine
Purity: 96%
CAS No.: 766-16-5
Molecular Formula: C6H6FN
Formula Weight: 111.12
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Refractive Index: 1.4715
Sensitivity: Air Sensitive
Appearance: Colorless to pale yellow
Boiling Point: 128-132°
Beilstein Reference: 107087
MDL No.: MFCD04114145
Formula:
C₆H₆FN MW: 111.12 g/mol Boiling Pt: 128…132 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD04114145 CAS Number: 766-16-5 |
Specification Test Results
Assay Minimum | 96 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...