5-Azaindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine
H64272-03
H64272-06
AAH64272-03EA
44.2
USD
AAH64272-03
AAH64272-06
AAH64272-14
5-Azaindole 98%
5-Azaindole
Purity: 98%
CAS No.: 271-34-1
Molecular Formula: C7H6N2
Formula Weight: 118.14
Melting Point: 104-108°
Beilstein Reference: 109684
MDL No.: MFCD00955936
Formula:
C₇H₆N₂ MW: 118.14 g/mol Melting Pt: 104…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00955936 CAS Number: 271-34-1 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...