5-Methyluridine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidine-2,4(1H,3H)-dione

H64259-14 H64259-22
AAH64259-14EA 80.8 USD
AAH64259-14 AAH64259-22
5-Methyluridine 99%
5-Methyluridine

Purity: 99% CAS No.: 1463-10-1 Molecular Formula: C10H14N2O6 Formula Weight: 258.23 Storage Temperature: 2°C to 8°C Physical Form: Solid Appearance: Off-white to white Melting Point: 183-184° Beilstein Reference: 90164 Optical Rotation: -8°(C=2 in water) MDL No.: MFCD00006535

Formula: C₁₀H₁₄N₂O₆
MW: 258.23 g/mol
Melting Pt: 183…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00006535
CAS Number: 1463-10-1
EINECS: 215-973-9

Order Now

Specification Test Results

Assay Minimum 99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR