3-Bromopicolinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Bromopyridine-2-carboxylic acid, 3-Bromo-2-pyridinecarboxylic acid
H64258-06
H64258-14
AAH64258-06EA
78.8
USD
AAH64258-06
AAH64258-14
3-Bromopicolinic acid 97%
3-Bromopicolinic acid
Purity: 97%
CAS No.: 30683-23-9
Molecular Formula: C6H4BrNO2
Formula Weight: 202.01
Physical Form: Solid
Density: 1.813
Melting Point: 141-144°
Beilstein Reference: 472171
MDL No.: MFCD01320380
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Melting Pt: 141…144 °C Density: 1.813 Storage Temperature: Ambient |
MDL Number:
MFCD01320380 CAS Number: 30683-23-9 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...