4-Bromopicolinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromopyridine-2-carboxylic acid
H64244-03
H64244-06
H64244-14
AAH64244-03EA
37.44
USD
AAH64244-03
AAH64244-06
AAH64244-14
4-Bromopicolinic acid 97%
4-Bromopicolinic acid
Purity: 97%
CAS No.: 30766-03-1
Molecular Formula: C6H4BrNO2
Formula Weight: 202.01
Melting Point: 177-179°
Beilstein Reference: 471891
MDL No.: MFCD03426665
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Melting Pt: 177…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426665 CAS Number: 30766-03-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...