4-Azaindole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1H-Pyrrolo[3,2-b]pyridine, 1H-Pyrrolo[2,3-e]pyridine
H64224-03
H64224-06
AAH64224-03EA
55.9
USD
AAH64224-03
AAH64224-06
4-Azaindole 97%
4-Azaindole
Purity: 97%
CAS No.: 272-49-1
Molecular Formula: C7H6N2
Formula Weight: 118.14
Physical Form: Solid
Melting Point: 125-127°
Beilstein Reference: 109683
MDL No.: MFCD00971977
Formula:
C₇H₆N₂ Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00971977 CAS Number: 272-49-1 |
Specification Test Results
Assay Minimum | 97 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...