3-Bromo-9-phenyl-9H-carbazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Bromo-9-phenylcarbazole
H64212-06
H64212-14
H64212-22
AAH64212-06EA
49.4
USD
AAH64212-06
AAH64212-14
AAH64212-22
3-Bromo-9-phenyl-9H-carbazole 98%
3-Bromo-9-phenyl-9H-carbazole
Purity: 98%
CAS No.: 1153-85-1
Molecular Formula: C18H12BrN
Formula Weight: 322.20
Physical Form: Solid
Melting Point: 97-98°
Beilstein Reference: 1315492
MDL No.: MFCD11977305
Formula:
C18H12BrN MW: 322.20 g/mol Melting Pt: 97…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD11977305 CAS Number: 1153-85-1 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...