1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%

Supplier: Thermo Scientific Chemicals
Warning

H64191-03 H64191-06
AAH64191-03EA 37.1 USD
AAH64191-03 AAH64191-06
1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%
1-Methyl-1,2,3,4-tetrahydroisoquinoline

Purity: 95% CAS No.: 4965-09-7 Molecular Formula: C10H13N Formula Weight: 147.22 Storage Temperature: -30°C to -10°C Physical Form: Liquid Appearance: Pale yellow Boiling Point: 119°/18mm Beilstein Reference: 82140 MDL No.: MFCD00798989

Formula: C10H13N
MW: 147.22 g/mol
Boiling Pt: 119 °C/18 mm
Storage Temperature: Refrigerator
MDL Number: MFCD00798989
CAS Number: 4965-09-7

Order Now

Specification Test Results

Assay Minimum 95

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR