4-(Trifluoromethyl)anisole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H64187-03
H64187-06
H64187-MD
AAH64187-03EA
61.6
USD
AAH64187-03
AAH64187-06
AAH64187-MD
4-(Trifluoromethyl)anisole 98%
4-(Trifluoromethyl)anisole
Purity: 98%
CAS No.: 402-52-8
Molecular Formula: C8H7F3O
Formula Weight: 176.14
Storage Temperature: 2°C to 8°C
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.245
Refractive Index: 1.4450
Appearance: Colorless to pale yellow
Melting Point: -9°
Boiling Point: 169°/754mm
UN No.: 1993
MDL No.: MFCD03094456
Formula:
C₈H₇F₃O MW: 176.14 g/mol Boiling Pt: 169 °C/754 mm Melting Pt: –9 °C Density: 1.245 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03094456 CAS Number: 402-52-8 UN: 1993 ADR: 3,III |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...