1-Boc-pyrrole-2-boronic acidpinacol ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: tert-Butyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrole-1-carboxylate

H64171-03 H64171-06
AAH64171-03EA 73.8 USD
AAH64171-03 AAH64171-06 AAH64171-MD
1-Boc-pyrrole-2-boronic acidpinacol ester 98%
1-Boc-pyrrole-2-boronic acidpinacol ester

Purity: 98% CAS No.: 1072944-98-9 Molecular Formula: C15H24BNO4 Formula Weight: 293.17 Storage Temperature: -30°C to -10°C Physical Form: Solid MDL No.: MFCD11504960

Formula: C₁₅H₂₄BNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD11504960
CAS Number: 1072944-98-9

Order Now

Specification Test Results

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR