2,5-Dimethoxy-4-methylbenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

H64140-03 H64140-06 H64140-14
AAH64140-03EA 56.5 USD
AAH64140-03 AAH64140-06 AAH64140-14
2,5-Dimethoxy-4-methylbenzaldehyde 97%
2,5-Dimethoxy-4-methylbenzaldehyde

Purity: 97% CAS No.: 4925-88-6 Molecular Formula: C10H12O3 Formula Weight: 180.20 Physical Form: Solid Appearance: Off-white Melting Point: 82-86° Beilstein Reference: 1866592 MDL No.: MFCD02253192

Formula: C₁₀H₁₂O₃
Melting Pt: 82…86 °C
Storage Temperature: Ambient
MDL Number: MFCD02253192
CAS Number: 4925-88-6

Order Now

Specification Test Results

Assay Minimum 97


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR